Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylidine Ponceau 2R, pure, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
3761-53-3
C18H14N2Na2O7S2
480.417
MFCD00003897
YJVBLROMQZEFPA-UHFFFAOYSA-L
4-(2,4-Dimethylphenylazo)-3-hydroxy-2,7-naphthalenedisulfonic acid, disodium salt, C.I. 16150, 4-(2, 4-Dimethylphenylazo)-3-hydroxy-2, 7-naphthalenedisulfonic acid, Ponceau G, R, 2R
102092631
CHEBI:82369
disodium;5-[(2,4-dimethylphenyl)diazenyl]-6-oxido-7-sulfonaphthalene-2-sulfonate
CC1=CC(=C(C=C1)N=NC2=C3C=CC(=CC3=CC(=C2[O-])S(=O)(=O)O)S(=O)(=O)[O-])C.[Na+].[Na+]
3761-53-3
C18H14N2Na2O7S2
480.417
MFCD00003897
YJVBLROMQZEFPA-UHFFFAOYSA-L
4-(2,4-Dimethylphenylazo)-3-hydroxy-2,7-naphthalenedisulfonic acid, disodium salt, C.I. 16150, 4-(2, 4-Dimethylphenylazo)-3-hydroxy-2, 7-naphthalenedisulfonic acid, Ponceau G, R, 2R
102092631
CHEBI:82369
disodium;5-[(2,4-dimethylphenyl)diazenyl]-6-oxido-7-sulfonaphthalene-2-sulfonate
CC1=CC(=C(C=C1)N=NC2=C3C=CC(=CC3=CC(=C2[O-])S(=O)(=O)O)S(=O)(=O)[O-])C.[Na+].[Na+]
Infrared Spectrum Authentic
Formula Weight 480.41
Physical Form Fine Powder
Grade Extra Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Bordeaux to Red
Quantity 100g
Chemical Name or Material Xylidine Ponceau 2R

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Suspected of causing cancer.
Causes serious eye irritation.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

CANCER SUSPECTED AGENT

EINECSNumber : 223-178-3

RTECSNumber : QJ6825000

TSCA : TSCA