Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylitol, NF, 98.5-101%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
87-99-0
C5H12O5
152.15
HEBKCHPVOIAQTA-UHFFFAOYNA-N
pentane-1,2,3,4,5-pentol
OCC(O)C(O)C(O)CO
87-99-0
C5H12O5
152.15
HEBKCHPVOIAQTA-UHFFFAOYNA-N
pentane-1,2,3,4,5-pentol
OCC(O)C(O)C(O)CO

X1017, 87-99-0, C5H12O5

Spectrum™ Xylitol, NF is used as a sugar substitute in pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Pail
Quantity 12 kg
Water 0.005
Ignition Residue 0.005
CAS Max % 1
Percent Purity 98.5 to 101%
Grade NF