Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylometazoline Hydrochloride, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
1218-35-5
C16H25ClN2
280.84
YGWFCQYETHJKNX-UHFFFAOYSA-N
hydrogen 2-[(4-tert-butyl-2,6-dimethylphenyl)methyl]-4,5-dihydro-1H-imidazole chloride
[H+].[Cl-].CC1=CC(=CC(C)=C1CC1=NCCN1)C(C)(C)C
1218-35-5
C16H25ClN2
280.84
YGWFCQYETHJKNX-UHFFFAOYSA-N
hydrogen 2-[(4-tert-butyl-2,6-dimethylphenyl)methyl]-4,5-dihydro-1H-imidazole chloride
[H+].[Cl-].CC1=CC(=CC(C)=C1CC1=NCCN1)C(C)(C)C

X2001, 1218-35-5, C16H24N2•HCl

Spectrum™ Xylometazoline Hydrochloride , also known as xylomethazoline, is used topically as a nasal decongestant in either drops or sprays. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Packaging Amber Glass Bottle
Quantity 100 g
CAS Max % 1
UN Number UN2811