Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylometazoline Hydrochloride, USP, 99-101%, Spectrum™

Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle

X2012, 1218-35-5, C16H24N2•HCl

Spectrum™ Xylometazoline Hydrochloride, USP is used to treat stuffy nose caused by allergies, sinus irritation, or the common cold. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Loss on Drying 0.005
Ignition Residue 0.001
Quantity 5 g
CAS 1218-35-5
CAS Max % 1
Molecular Formula C16H25ClN2
InChI Key YGWFCQYETHJKNX-UHFFFAOYSA-N
SMILES [H+].[Cl-].CC1=CC(=CC(C)=C1CC1=NCCN1)C(C)(C)C
IUPAC Name hydrogen 2-[(4-tert-butyl-2,6-dimethylphenyl)methyl]-4,5-dihydro-1H-imidazole chloride
Molecular Weight (g/mol) 280.84
Percent Purity 99 to 101%
Grade USP
Show More Show Less