Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylometazoline Hydrochloride, USP, 99-101%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
Packaging:
Amber Glass Bottle
1218-35-5
C16H25ClN2
280.84
YGWFCQYETHJKNX-UHFFFAOYSA-N
hydrogen 2-[(4-tert-butyl-2,6-dimethylphenyl)methyl]-4,5-dihydro-1H-imidazole chloride
[H+].[Cl-].CC1=CC(=CC(C)=C1CC1=NCCN1)C(C)(C)C
1218-35-5
C16H25ClN2
280.84
YGWFCQYETHJKNX-UHFFFAOYSA-N
hydrogen 2-[(4-tert-butyl-2,6-dimethylphenyl)methyl]-4,5-dihydro-1H-imidazole chloride
[H+].[Cl-].CC1=CC(=CC(C)=C1CC1=NCCN1)C(C)(C)C

X2012, 1218-35-5, C16H24N2•HCl

Spectrum™ Xylometazoline Hydrochloride, USP is used to treat stuffy nose caused by allergies, sinus irritation, or the common cold. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Ignition Residue 0.001
Loss on Drying 0.005
Quantity 100 g
CAS Max % 1
Percent Purity 99 to 101%
Grade USP