Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylose, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
25 g
5 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
58-86-6
C5H10O5
150.13
SRBFZHDQGSBBOR-UHFFFAOYNA-N
oxane-2,3,4,5-tetrol
OC1COC(O)C(O)C1O
58-86-6
C5H10O5
150.13
SRBFZHDQGSBBOR-UHFFFAOYNA-N
oxane-2,3,4,5-tetrol
OC1COC(O)C(O)C1O

XY110, 58-86-6, C5H10O5

Spectrum™ Xylose, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Pail
Ignition Residue 0.0005
Loss on Drying 0.001
Specific Rotation +18.2° to +19.4°
Quantity 5 kg
CAS Max % 1
Percent Purity 98 to 102%
Grade USP