Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Y 11, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
1086639-59-9
C8H17BrN4O
265.155
HBPXFHNNLMCUPA-UHFFFAOYSA-M
1-2-hydroxyethyl-1,3,5,7-tetraazatricyclo 3.3.1.1 3 ,? decan-1-ium bromide,1-2-hydroxyethyl-3,5,7-triaza-1-azaniatricyclo 3.3.1.13,7 decane bromide,3,7-triaza-1-azoniatricyclo 3.3.1.13,7 decane, 1-2-hydroxyethyl-, bromide
24195918
C1N2CN3CN1C[N+](C2)(C3)CCO.[Br-]
This item is not returnable. View return policy
1086639-59-9
C8H17BrN4O
265.155
HBPXFHNNLMCUPA-UHFFFAOYSA-M
1-2-hydroxyethyl-1,3,5,7-tetraazatricyclo 3.3.1.1 3 ,? decan-1-ium bromide,1-2-hydroxyethyl-3,5,7-triaza-1-azaniatricyclo 3.3.1.13,7 decane bromide,3,7-triaza-1-azoniatricyclo 3.3.1.13,7 decane, 1-2-hydroxyethyl-, bromide
24195918
C1N2CN3CN1C[N+](C2)(C3)CCO.[Br-]

Potent and selective FAK inhibitor

Potent inhibitor of focal adhesion kinase (FAK); prevents FAK autophosphorylation at the Y397 site (IC50 ∽ 50 nM in an in vitro kinase assay). Displays selectivity for FAK over a panel of other kinases (concentration used in assay = 1 μM). Decreases cell viability in a number of cancer cell lines; blocks colony formation in SW620 and BT474 cell lines.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 265.15
Percent Purity >98%
Chemical Name or Material Y 11

Recommended Storage : Store at -20°C