Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Y-26763, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Y-26763 is a Kir6 (KATP) channel opener and active metabolite of Y-27152. Relaxes contracted rat aortic rings (IC50 = 0.027 mM) and inhibits glucose-induced insulin secretion in isolated human pancreatic β-cells in vitro. Produces hypotension in spontaneously hypertensive rats and displays cardioprotective properties in dogs following systemic administration in vivo.
Specifications
Quantity | 10 mg |
Formula Weight | 276.29 |
Percent Purity | >98% |
Chemical Name or Material | Y-26763 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction