Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Y-27152, Tocris Bioscience™

Catalog Number 207750
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
127408-30-4
C21H22N2O4
366.417
RHFUXPCCELGMFC-UHFFFAOYSA-N
acmc-20msfk, acmc-20n5s4, acetamide,n-6-cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2h-1-benzopyran-4-yl-n-phenylmethoxy-,trans-9ci, acetamide,n-3s,4r-6-cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2h-1-benzopyran-4-yl-n-phenylmethoxy
19042657
N-(6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydrochromen-4-yl)-N-phenylmethoxyacetamide
CC(=O)N(C1C(C(OC2=C1C=C(C=C2)C#N)(C)C)O)OCC3=CC=CC=C3
This item is not returnable. View return policy
127408-30-4
C21H22N2O4
366.417
RHFUXPCCELGMFC-UHFFFAOYSA-N
acmc-20msfk, acmc-20n5s4, acetamide,n-6-cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2h-1-benzopyran-4-yl-n-phenylmethoxy-,trans-9ci, acetamide,n-3s,4r-6-cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2h-1-benzopyran-4-yl-n-phenylmethoxy
19042657
N-(6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydrochromen-4-yl)-N-phenylmethoxyacetamide
CC(=O)N(C1C(C(OC2=C1C=C(C=C2)C#N)(C)C)O)OCC3=CC=CC=C3

Prodrug of Kir6 (KATP) channel opener Y-26763 (Cat. No. 2076); orally active in vivo

Prodrug of the Kir6 (KATP) channel opener Y-26763 (Cat. No. 2076). Inactive in vitro; fails to modify tension in contracted aortic rings at concentrations up to 1 mM. Produces long-lasting antihypertensive effects in vivo, with minimal tachycardia following oral administration in hypertensive animals.
Quantity 50 mg
Formula Weight 366.41
Percent Purity >98%
Chemical Name or Material Y-27152
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit