Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Y-29794 oxalate, Tocris Bioscience™

Catalog Number 163450
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
146794-84-5
C25H36N2O5S2
508.692
UUSSHPPSOUAHHP-UHFFFAOYSA-N
y-29794 oxalate, 8-6-isopropyl-3-thiophene-2-carbonyl pyridin-2-yl sulfanyl octyl dimethylamine; oxalic acid, 2-8-dimethylamino octyl thio-6-1-methylethyl-3-pyridinyl-2-thienylmethanone oxalate
45073463
[2-[8-(dimethylamino)octylsulfanyl]-6-propan-2-ylpyridin-3-yl]-thiophen-2-ylmethanone;oxalic acid
CC(C)C1=NC(=C(C=C1)C(=O)C2=CC=CS2)SCCCCCCCCN(C)C.C(=O)(C(=O)O)O
This item is not returnable. View return policy
146794-84-5
C25H36N2O5S2
508.692
UUSSHPPSOUAHHP-UHFFFAOYSA-N
y-29794 oxalate, 8-6-isopropyl-3-thiophene-2-carbonyl pyridin-2-yl sulfanyl octyl dimethylamine; oxalic acid, 2-8-dimethylamino octyl thio-6-1-methylethyl-3-pyridinyl-2-thienylmethanone oxalate
45073463
[2-[8-(dimethylamino)octylsulfanyl]-6-propan-2-ylpyridin-3-yl]-thiophen-2-ylmethanone;oxalic acid
CC(C)C1=NC(=C(C=C1)C(=O)C2=CC=CS2)SCCCCCCCCN(C)C.C(=O)(C(=O)O)O

Prolyl endopeptidase inhibitor

Inhibitor of prolyl endopeptidase, a serine peptidase that may be implicated in the biosynthesis of amyloid β-peptide. Potently inhibits rat brain prolyl endopeptidase (Ki = 0.95 nM) in vitro, and prevents amyloid β-peptide deposition in vivo. Orally active and brain penetrant.
Quantity 50 mg
Formula Weight 508.69
Percent Purity >98%
Chemical Name or Material Y-29794 oxalate
Recommended Storage Desiccate at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit