Learn More
Cole-Parmer Masterflex Fitting, PVDF, Y-Connector, Hose Barb Union, 3/8" ID; 100/PK  DFS Item
Manufacturer:  Cole-Parmer 3072766
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications. Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing. The PVDF fittings offer excellent chemical resistance. Temp range: -80 to 265°F (-62 to 129°
Catalog No. 50-630-773