Learn More
Cole-Parmer Masterflex Fitting, Nylon, Y-Connector, Hose Barb Union, 1/2" ID; 10/PK  DFS Item
Manufacturer:  Cole-Parmer 3061422
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications. Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing. These nylon fittings offer good chemical resistance. Temp range: -50 to 250°F (-46 to 121°C). Sterilize by ethylene oxide only.
Catalog No. 50-630-811