Learn More
Cole-Parmer Masterflex Fitting, HDPE, Y-Connector, Hose Barb Union, 1/4" ID; 10/PK  DFS Item
Manufacturer:  Cole-Parmer 3061413
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications. Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing. These High-Density Polyethylene (HDPE) fittings offer very good chemical resistance. Temp range: -65 to 150°F (-53 to 65°C). Sterilize by ethylene oxide only.
Catalog No. 50-630-762