Learn More
Fisherbrand™ Y-Shaped Connectors with Straight Ends Products for Science Education
Autoclavable and highly resistant to many chemicals.
 View more versions of this product
Catalog No. S37185
Add to cart
Description
Description
Specifications
Specifications
Transparent
Autoclavable
15.9 mm
3.8 in.
Tubing Connector
16.2 mm
Y
2.35 in.
20 psi
0.625 in.
Polypropylene
96.5 mm
16 mm
0.64 in.
59.6 mm