Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YAP1 Mouse anti-Human, Alexa Fluor™ 532, Clone: 1A12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222117AF532

Catalog No. NP222117A32


Add to cart

Description

Description

YAP1 Monoclonal antibody specifically detects YAP1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

YAP1
1A12
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
YAP1
Purified recombinant fragment of human YAP1 expressed in E. coli. [UniProtNo. P46937]
Ammonium sulfate precipitation
Breast Cancer, Cancer, Chromatin Research, Core ESC Like Genes, Phospho Specific, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators, Tyrosine Kinases
Primary
10413.0
ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
65 kDa Yes-associated protein, YAP, YAP2, YAP65yes-associated protein 2, Yes-associated protein 1, Yes-associated protein 1, 65kDa, yes-associated protein beta, yes-associated protein delta, YKI, yorkie homolog
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit