Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YARS Polyclonal Antibody
YARS Polyclonal Antibody
Supplier: Thermo Scientific PA527434
Description
YARS Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Specifications
YARS | |
Polyclonal | |
Unconjugated | |
YARS1 | |
AL024047; AU018965; CMTDIC; tyrosine tRNA ligase 1, cytoplasmic; tyrosine--tRNA ligase, cytoplasmic; Tyrosine--tRNA ligase, cytoplasmic, N-terminally processed; tyrosyl trna synthetase; tyrosyl--tRNA ligase; tyrosyl-tRNA synthetase; tyrosyl-tRNA synthetase, cytoplasmic; tyrRS; Yars; YRS; YTS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
107271, 8565 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.25 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P54577, Q91WQ3 | |
YARS1 | |
Recombinant fragment corresponding to a region within amino acids 307 and 528 of Human Tyrosyl tRNA synthetase. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction