Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YARS2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550344

Catalog No. PA550344


Add to cart

Description

Description

The antibody detects endogenous levels of total YARS2 protein.

This gene encodes a mitochondrial protein that catalyzes the attachment of tyrosine to tRNA(Tyr). Mutations in this gene are associated with myopathy with lactic acidosis and sideroblastic anemia type 2 (MLASA2).
Specifications

Specifications

YARS2
Polyclonal
Unconjugated
YARS2
Liquid
YARS2
Fusion protein corresponding to residues near the C terminal of human tyrosyl-tRNA synthetase 2, mitochondrial
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q9Y2Z4, Q8BYL4
TYRRS, CGI-04, MLASA2, MT-TYRRS
Rabbit
IgG
100 μL
-20°C
Primary
51067, 70120
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit