Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YB1 Mouse anti-Human, Clone: OTI7B4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274905

Catalog No. NBP274905


Add to cart

Description

Description

YB1 Monoclonal antibody specifically detects YB1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

YB1
OTI7B4
Unconjugated
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
BP-8, CBF-A, class II, Y box-binding protein I, CSDA2, CSDB, DBPB CCAAT-binding transcription factor I subunit A, EFI-A, Enhancer factor I subunit A, MDR-NF1, MGC104858, MGC110976, MGC117250, NSEP1 Y-box-binding protein 1, nuclease-sensitive element-binding protein 1, Y box binding protein 1, YB1 DNA-binding protein B, YB-1 Y-box transcription factor, YBX1
Mouse
IgG1
Immunogen affinity purified
Breast Cancer, Cancer, DNA Repair, Epigenetics, Ovarian Carcinoma Cell Markers, Stem Cell Markers, Transcription Factors and Regulators, Translation Control
Primary
4904
Human
Immunohistochemistry, Western Blot
LYOPH
Western Blot 1:2000, Immunohistochemistry 1:150
Affinity Purified, Lyophilized
YBX1
Full length human recombinant protein of human YBX1 (NP_004550) produced in E.coli.
35.7 kDa
100 μg
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit