Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YB1 Mouse anti-Human, Clone: rYBX1/2430, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30746110UG

 View more versions of this product

Catalog No. NB122395


Add to cart

Description

Description

YB1 Monoclonal specifically detects YB1 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

YB1
rYBX1/2430
Unconjugated
10 mM PBS with 0.05% BSA
BP-8, CBF-A, class II, Y box-binding protein I, CSDA2, CSDB, DBPB CCAAT-binding transcription factor I subunit A, EFI-A, Enhancer factor I subunit A, MDR-NF1, MGC104858, MGC110976, MGC117250, NSEP1 Y-box-binding protein 1, nuclease-sensitive element-binding protein 1, Y box binding protein 1, YB1 DNA-binding protein B, YB-1 Y-box transcription factor, YBX1
Recombinant human full-length YB1 protein
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Immunohistochemistry (Paraffin)
0.2 mg/ml
Immunohistochemistry-Paraffin 1-2 ug/ml
Purified
Mouse
IgG1 Kappa
100 μg
Breast Cancer, Cancer, DNA Repair, Epigenetics, Ovarian Carcinoma Cell Markers, Stem Cell Markers, Transcription Factors and Regulators, Translation Control
Primary
4904
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit