Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YBX2 Mouse anti-Human, Mouse, Clone: OTI3B5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527431

Catalog No. PIMA527431


Add to cart

Description

Description

Germ cell specific Y-box binding protein (YBX2), a germ-cell-specific member of the Y-box family of DNA-/RNA-binding proteins, is a major constituent of messenger ribonucleoprotein particles (mRNPs) and involved in the regulation of the stability and/or translation of germ cell mRNAs. It is proposed to function as a coactivator of transcription in the nucleus and to stabilize and store maternal and paternal mRNAs in the cytoplasm. YBX2 binds to the Y-box consensus promoter element and is expressed in oocytes and testicular germ cells in the stage of spermatogonia to spermatocyte. Recent studies show that deletion of YBX2 leads to the disruption of both spermatogenesis and oogenesis.
Specifications

Specifications

YBX2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y2T7, Q9Z2C8
CONTRIN, CSDA3, DBPC, MSY2; DNA-binding protein C; MSY2 homolog; Y-box-binding protein 2; germ cell specific Y-box binding protein; germ cell-specific Y-box-binding protein
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51087, 53422
Immunohistochemistry (Paraffin), Western Blot
OTI3B5
Unconjugated
YBX2
Liquid
YBX2
Full length human Recombit protein of YBX2 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit