Learn More
Medchemexpress LLC Yeast extract | 8013-01-2 | 50 G
SDP
Supplier:  Medchemexpress LLC HY15312650G
Encompass_Preferred
Yeast extract, derived from *Saccharomyces cerevisiae*, is a concentrate of soluble yeast components. It is a rich source of nutritional elements including partially hydrolyzed proteins, free amino acids, B vitamins, and minerals. This versatile ingredient offers both nutritional and flavoring properties, and research suggests it can promote wound healing.
  • Rich in nutritional components, including partially hydrolyzed proteins, free amino acids, B vitamins, and minerals
  • Acts as a food ingredient for nutritional and flavoring purposes
  • Promotes wound healing in various models
  • Supports rapid growth and cell viability in specific Rhizobium species
  • Accelerates wound closure and cellular reorganization in diabetic mice wound models
Catalog No. 50-003-73153