Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Yeast FCY1 (NP_015387.1, 1 a.a - 158 a.a.) Full length Recombinant Protein with His tag

Used for Func, SDS-PAGE

Manufacturer:  Abnova™ P6203

Catalog No. 89-944-354


Add to cart

Specifications

Specifications

NP_015387.1
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
77.5kDa
Escherichia coli expression system
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
FCY1
Escherichia coli expression system
Escherichia coli
His
2 U/uL
Liquid
856175
FCY1 (Yeast) Recombinant Protein
100 Units
RUO
FCY1
1U FCY1 converts > 80% of 10 mg cytosine to uracil at 37°C in 90 minutes.
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit