Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Yeast HSP104 (NP_013074, 1 a.a. - 908 a.a.) Full-length Recombinant Protein

Used for SDS-PAGE

Manufacturer:  Abnova™ P4985

Catalog No. 89-948-019


Add to cart

Description

Description

Sequence: MNDQTQFTERALTILTLAQKLASDHQHPQLQPIHILAAFIETPEDGSVPYLQNLIEKGRYDYDLFKKVVNRNLVRIPQQQPAPAEITPSYALGKVLQDAAKIQKQQKDSFIAQDHILFALFNDSSIQQIFKEAQVDIEAIKQQALELRGNTRIDSRGADTNTPLEYLSKYAIDMTEQARQGKLDPVIGREEEIRSTIRVLARRIKSNPCLIGEPGIGKTAIIEGVAQRIIDDDVPTILQGAKLFSLDLAALTAGAKYKGDFEERFKGVLKEIEESKTLIVLFIDEIHMLMGNGKDDAANILKPALSRGQLKVIGATTNNEYRSIVEKDGAFERRFQKIEVAEPSVRQTVAILRGLQPKYEIHHGVRILDSALVTAAQLAKRYLPYRRLPDSALDLVDISCAGVAVARDSKPEELDSKERQLQLIQVEIKALERDEDADSTTKDRLKLARQKEASLQEELEPLRQRYNEEKHGHEELTQAKKKLDELENKALDAERRYDTATAADLRYFAIPDIKKQIEKLEDQVAEEERRAGANSMIQNVVDSDTISETAARLTGIPVKKLSESENEKLIHMERDLSSEVVGQMDAIKAVSNAVRLSRSGLANPRQPASFLFLGLSGSGKTELAKKVAGFLFNDEDMMIRVDCSELSEKYAVSKLLGTTAGYVGYDEGGFLTNQLQYKPYSVLLFDEVEKAHPDVLTVMLQMLDDGRITSGQGKTIDCSNCIVIMTSNLGAEFINSQQGSKIQESTKNLVMGAVRQHFRPEFLNRISSIVIFNKLSRKAIHKIVDIRLKEIEERFEQNDKHYKLNLTQEAKDFLAKYGYSDDMGARPLNRLIQNEILNKLALRILKNEIKDKETVNVVLKKGKSRDENVPEEAEECLEVLPNHEATIGADTLGDDDNEDSMEIDDDLD
Specifications

Specifications

Escherichia coli expression system
1 mg/mL
Liquid
850633
HSP104 (Yeast) Recombinant Protein
Conventional Chromatography
Loading 3 ug protein in 10% SDS-PAGE
MNDQTQFTERALTILTLAQKLASDHQHPQLQPIHILAAFIETPEDGSVPYLQNLIEKGRYDYDLFKKVVNRNLVRIPQQQPAPAEITPSYALGKVLQDAAKIQKQQKDSFIAQDHILFALFNDSSIQQIFKEAQVDIEAIKQQALELRGNTRIDSRGADTNTPLEYLSKYAIDMTEQARQGKLDPVIGREEEIRSTIRVLARRIKSNPCLIGEPGIGKTAIIEGVAQRIIDDDVPTILQGAKLFSLDLAALTAGAKYKGDFEERFKGVLKEIEESKTLIVLFIDEIHMLMGNGKDDAANILKPALSRGQLKVIGATTNNEYRSIVEKDGAFERRFQKIEVAEPSVRQTVAILRGLQPKYEIHHGVRILDSALVTAAQLAKRYLPYRRLPDSALDLVDISCAGVAVARDSKPEELDSKERQLQLIQVEIKALERDEDADSTTKDRLKLARQKEASLQEELEPLRQRYNEEKHGHEELTQAKKKLDELENKALDAERRYDTATAADLRYFAIPDIKKQIEKLEDQVAEEERRAGANSMIQNVVDSDTISETAARLTGIPVKKLSESENEKLIHMERDLSSEVVGQMDAIKAVSNAVRLSRSGLANPRQPASFLFLGLSGSGKTELAKKVAGFLFNDEDMMIRVDCSELSEKYAVSKLLGTTAGYVGYDEGGFLTNQLQYKPYSVLLFDEVEKAHPDVLTVMLQMLDDGRITSGQGKTIDCSNCIVIMTSNLGAEFINSQQGSKIQESTKNLVMGAVRQHFRPEFLNRISSIVIFNKLSRKAIHKIVDIRLKEIEERFEQNDKHYKLNLTQEAKDFLAKYGYSDDMGARPLNRLIQNEILNKLALRILKNEIKDKETVNVVLKKGKSRDENVPEEAEECLEVLPNHEATIGADTLGDDDNEDSMEIDDDLD
RUO
HSP104
Yes
NP_013074
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
102kDa
Escherichia coli expression system
>90% by SDS-PAGE
100 ug
Store at 4°C short term (1-2 weeks). For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
HSP104
Escherichia coli
None
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit