Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Yeast Kex-2 Recombinant Protein, PeproTech®
Insect cells Recombinant Protein
Specifications
Accession Number | P13134 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 855483 |
Molecular Weight (g/mol) | 60.4 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
45045250UG
|
Gibco™
45045250UG |
250 μg |
Each for $217.00
|
N/A | |||||
45045500UG
|
Gibco™
45045500UG |
500 μg |
Each for $400.00
|
N/A
|
|||||
450-45-50UG
|
Gibco™
4504550UG |
50 μg |
Each for $89.50
|
N/A
|
|||||
450-45-1MG
|
Gibco™
450451MG |
2 x 500 μg | N/A | N/A | N/A | ||||
Description
Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides. Most proteases act in a specific manner, hydrolyzing bonds at, or adjacent to, specific residues, or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes, including prenatal and postnatal development, reproduction, signal transduction, immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, as they are frequently used in the analysis and production of proteins. Kex-2 cleaves at the carboxyl end of the recognition sequences Arg-Arg/X and Lys-Arg/X.Specifications
P13134 | |
protein with no preservative | |
60.4 | |
Insect cells | |
RUO | |
KEX2 | |
Unconjugated | |
LPVPAPPMDS SLLPVKEAED KLSINDPLFE RQWHLVNPSF PGSDINVLDL WYNNITGAGV VAAIVDDGLD YENEDLKDNF CAEGSWDFND NTNLPKPRLS DDYHGTRCAG EIAAKKGNNF CGVGVGYNAK ISGIRILSGD ITTEDEAASL IYGLDVNDIY SCSWGPADDG RHLQGPSDLV KKALVKGVTE GRDSKGAIYV FASGNGGTRG DNCNYDGYTN SIYSITIGAI DHKDLHPPYS EGCSAVMAVT YSSGSGEYIH SSDINGRCSN SHGGTSAAAP LAAGVYTLLL EANPNLTWRD VQYLSILSAV GLEKNADGDW RDSAMGKKYS HRYGFGKIDA HKLIEMSKTW ENVNAQTWFY LPTLYVSQST NSTEETLESV ITISEKSLQD ANFKRIEHVT VTVDIDTEIR GTTTVDLISP AGIISNLGVV RPRDVSSEGF KDWTFMSVAH WGENGVGDWK IKVKTTENGH RIDFHSWRLK LFGESIDSSK TETFVFGNDK EEVEPAATES TVSQYSASST SISISATSTS SISIGVETSA IPQTTTASTD PDSDPNTP | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
855483 | |
Yeast Kex-2 | |
-20°C | |
<1 EU/μg | |
Yeast | |
Recombinant | |
Lyophilized |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title