Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yeast Nitrogen Base, without amino acids, without ammonium sulfate, Alfa Aesar™

Used for classifying yeasts based on carbohydrate and amino acids requirements. Recommended for use in classifying yeasts based on carbon and nitrogen requirements. Contains all essential nutrients required for the growth of yeasts except amino acids, nitrogen and carbohydrate.

$179.40 - $550.50

Specifications

Packaging Plastic Bottle
Format Powder
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAH2627136 View Documents Alfa Aesar
H2627136
500g Each for $550.50
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAH2627122 View Documents Alfa Aesar
H2627122
100g Each for $179.40
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

Plastic Bottle
Powder
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit