Learn More
BioVision IncSupplier Diversity Partner Yeast Nuclei Isolation Kit: Yeast Nuclei Isolation Kit: Rapid & convenient kit for isolating yeast nuclei. High yield and purity. 50 Assays DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K28950
To understand the nature of the replication and expression of the yeast genome, it is essential to have a method for the preparation of nuclei at different stages of growth. BioVision’s Yeast Nuclei Isolation kit enables fast and easy purification of nuclei from yeast cells, utilizing yeast cell wall lysis and homogenization. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9670