Learn More
Brady™ Yellow Metal Write-On Tag
Identify facility equipment to your exact specifications. Brady™ Yellow Metal Write-On Tags are durable one-sided tags with a write-on area that accepts most marking pens.
Manufacturer:  Brady™ 56936
Catalog No. 19-113-069
Add to cart
Description
Description
  • Metal write-on tags are made a durable yet flexible metal material that can be bent without cracking
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor or outdoor use
  • Tags are cracking-resistant resistant
Specifications
Specifications
Yellow
Nylon fastener
1/Tag
Metal
3 in. x 5 in.
Not laminated
Rectangle