Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Yes Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems MAB32051SP
This item is not returnable.
View return policy
Description
Yes Monoclonal specifically detects Yes in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Yes | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunohistochemistry 8-25 ug/mL | |
P07947.3 | |
YES1 | |
E. coli-derived recombinant human Yes Met1-Gly88 Accession # P07947.3 | |
25 μg | |
Primary | |
Detects endogenous human, mouse, and rat Yes in Western blots. | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
339827 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
cellular yes-1 protein, c-yes, EC 2.7.10, EC 2.7.10.2, HsT441, P61-YES, Proto-oncogene c-Yes, proto-oncogene tyrosine-protein kinase Yes, v-yes-1 Yamaguchi sarcoma viral oncogene homolog 1, Yamaguchi sarcoma oncogene, Yes, YES1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7525 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction