Learn More
YES1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 202431AP
Catalog No. 50-173-2789
Add to cart
Description
Description
The antibody is specific to YES1.
YES is a member of the Src family of non-receptor tyrosine kinases participating in a wide range of cellular signalling pathways. The gene that encodes YES is the cellular homolog of the Yamaguchi sarcoma virus oncogene. This gene lies in proximity to thymidylate synthase gene on chromosome 18, and a pseudogene of it has mapped to chromosome 22.
Specifications
Specifications
YES1
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
F1LM93, P07947, Q04736
c yes, HsT441, P61 YES, Proto oncogene c Yes, Yes, YES1
Rabbit
IgG
150 μL
-20°C
Primary
22612, 24884, 7525
Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
YES1
Liquid
YES1
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat