Learn More
YIL 781, Tocris™
Ghrelin receptor (GHS-R1a) antagonist
Manufacturer:  R&D Systems 3959/50
Quantity 50mg
 View more versions of this product
Catalog No. 39-595-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
445.96
YIL 781
C24H28FN3O2·HCl
6-(4-Fluorophenoxy)-2-methyl-3-[[(3S)-1-(1-methylethyl)-3-piperidinyl]methyl]-4(3H)-quinazolinone
Specifications
50mg
>98%
445.96
YIL 781
875258-85-8
C24H28FN3O2·HCl
6-(4-Fluorophenoxy)-2-methyl-3-[[(3S)-1-(1-methylethyl)-3-piperidinyl]methyl]-4(3H)-quinazolinone
Safety and Handling
Recommended Storage : Desiccate at Room Temperature