Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YIPF3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548810

Catalog No. PA548810

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Involved in the maintece of the Golgi structure. May play a role in hematopoiesis.
Specifications

Specifications

YIPF3
Polyclonal
PBS with 0.09% sodium azide
Q9GZM5, Q3UDR8
Protein YIPF3, Killer lineage protein 1, Natural killer cell-specific antigen KLIP1, YIP1 family member 3, Protein YIPF3, 36 kDa form III, YIPF3, C6orf109, KLIP1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
25844, 28064
Western Blot
Unconjugated
YIPF3
Liquid
YIPF3
KLH conjugated synthetic peptide between 292-321 amino acids from the C-terminal region of human YIPF3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit