Learn More
Tocris Bioscience™ YK 3-237 Available on GSA/VA Contract for Federal Government customers only.
SIRT1 activator
$132.80 - $559.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
56-671-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5667/10
10mg Each for $132.80
Add to cart
56-675-0 View Documents Tocris Bioscience™
5667/50
Each for $559.00
Add to cart
Description
Description
SIRT1 activator. Inhibits proliferation of breast cancer cell lines expressing mutant p53. Induces G2/M cell cycle arrest and apoptosis in triple negative breast cancer cells (TNBCs) in vitro.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Safety and Handling
Safety and Handling
Recommended Storage : Store at −20°C