Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YM 202074, Tocris™

High affinity, selective mGlu1 antagonist

Manufacturer:  R&D Systems 3413/50

Quantity 50mg

 View more versions of this product

Catalog No. 34-135-0


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

>99%
588.67
YM 202074
C22H30N4O2S·1.5C4H4O4
N-Cyclohexyl-6-[[N-(2-methoxyethyl)-N-methylamino]methyl]-N-methylthiazolo[3, 2-a]benzoimidazole-2-carboxamide sesquifumarate

Specifications

50mg
>99%
588.67
YM 202074
299900-84-8
C22H30N4O2S·1.5C4H4O4
N-Cyclohexyl-6-[[N-(2-methoxyethyl)-N-methylamino]methyl]-N-methylthiazolo[3, 2-a]benzoimidazole-2-carboxamide sesquifumarate

Safety and Handling

Recommended Storage : Store at +4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit