Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YM 230888, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
446257-23-4
C19H28N4OS
360.52
SASNRPJKZVJWQW-OAHLLOKOSA-N
4-cycloheptylamino-n-2r-tetrahydro-2-furanyl methyl-thieno 2,3-d pyrimidine-6-methanamine,n-cycloheptyl-6-2r-tetrahydrofuran-2-ylmethyl amino methyl thieno 2,3-d pyrimidin-4-amine
11682007
N-cycloheptyl-6-[[[(2R)-oxolan-2-yl]methylamino]methyl]thieno[2,3-d]pyrimidin-4-amine
C1CCCC(CC1)NC2=C3C=C(SC3=NC=N2)CNCC4CCCO4
This item is not returnable. View return policy
446257-23-4
C19H28N4OS
360.52
SASNRPJKZVJWQW-OAHLLOKOSA-N
4-cycloheptylamino-n-2r-tetrahydro-2-furanyl methyl-thieno 2,3-d pyrimidine-6-methanamine,n-cycloheptyl-6-2r-tetrahydrofuran-2-ylmethyl amino methyl thieno 2,3-d pyrimidin-4-amine
11682007
N-cycloheptyl-6-[[[(2R)-oxolan-2-yl]methylamino]methyl]thieno[2,3-d]pyrimidin-4-amine
C1CCCC(CC1)NC2=C3C=C(SC3=NC=N2)CNCC4CCCO4

Selective mGlu1 antagonist

Selective mGlu 1 antagonist (K i = 13 nM). Inhibits mGlu 1 -mediated inositol phosphate production in rat cerebellar granule cells (IC 50 = 13 nM). Displays antinociceptive and analgesic effects in vitro; exhibits no significant sedative effect on locomotor activity.
Quantity 10mg
Formula Weight 360.52
Percent Purity >99%
Chemical Name or Material YM 230888

Recommended Storage : Store at Room Temperature