Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YM 90K hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
154164-30-4
C11H8ClN5O4
309.666
UMLFDVOHVJPDIZ-UHFFFAOYSA-N
ym 90k hydrochloride,ym 90k,ym-90k,uaexclwi0q,ym-90k hydrochloride,unii-uaexclwi0q,6-1h-imidazol-1-yl-7-nitro-2,3 1h,4h-quinoxalinedione,1,4-dihydro-6-1h-imidazol-1-yl-7-nitro-2,3-quinoxalinedione monohydrochloride,2,3-quinoxalinedione, 1,4-dihydro-6-1h-imidazol-1-yl-7-nitro-, monohydrochloride,ym90k
5486547
6-imidazol-1-yl-7-nitro-1,4-dihydroquinoxaline-2,3-dione;hydrochloride
C1=CN(C=N1)C2=C(C=C3C(=C2)NC(=O)C(=O)N3)[N+](=O)[O-].Cl
This item is not returnable. View return policy
154164-30-4
C11H8ClN5O4
309.666
UMLFDVOHVJPDIZ-UHFFFAOYSA-N
ym 90k hydrochloride,ym 90k,ym-90k,uaexclwi0q,ym-90k hydrochloride,unii-uaexclwi0q,6-1h-imidazol-1-yl-7-nitro-2,3 1h,4h-quinoxalinedione,1,4-dihydro-6-1h-imidazol-1-yl-7-nitro-2,3-quinoxalinedione monohydrochloride,2,3-quinoxalinedione, 1,4-dihydro-6-1h-imidazol-1-yl-7-nitro-, monohydrochloride,ym90k
5486547
6-imidazol-1-yl-7-nitro-1,4-dihydroquinoxaline-2,3-dione;hydrochloride
C1=CN(C=N1)C2=C(C=C3C(=C2)NC(=O)C(=O)N3)[N+](=O)[O-].Cl

Selective AMPA antagonist

Selective AMPA receptor antagonist (K i values are 84, 2200 and > 37000 nM for AMPA, kainate and NMDA receptors respectively). Neuroprotective; delays neuronal death in a global ischemia model and cerebral infarction in a focal ischemia model following postischemic administration.
Quantity 50mg
Formula Weight 309.67
Percent Purity >99%
Chemical Name or Material YM 90K hydrochloride

Recommended Storage : Desiccate at Room Temperature