Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YM 90K hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Selective AMPA receptor antagonist (Ki values are 84, 2200 and > 37000 nM for AMPA, kainate and NMDA receptors respectively). Neuroprotective; delays neuronal death in a global ischemia model and cerebral infarction in a focal ischemia model following postischemic administration.
Specifications
Quantity | 50 mg |
Formula Weight | 309.67 |
Percent Purity | >99% |
Chemical Name or Material | YM 90K hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction