Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YM 976, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
191219-80-4
C17H16ClN3O
313.785
MNHXYNNKDDXKNP-UHFFFAOYSA-N
4-3-chlorophenyl-1,7-diethylpyrido 2,3-d pyrimidin-2 1h-one,4-3-chlorophenyl-1,7-diethylpyrido 2,3-d pyrimidin-2-one,tocris-1821,d0c7jt,ym hplc,4-3-chlorophenyl-1,7-diethyl-1h,2h-pyrido 2,3-d pyrimidin-2-one,pyrido 2,3-d pyrimidin-2 1h-one,4-3-chlorophenyl-1,7-diethyl
6604918
4-(3-chlorophenyl)-1,7-diethylpyrido[2,3-d]pyrimidin-2-one
CCC1=NC2=C(C=C1)C(=NC(=O)N2CC)C3=CC(=CC=C3)Cl
This item is not returnable. View return policy
191219-80-4
C17H16ClN3O
313.785
MNHXYNNKDDXKNP-UHFFFAOYSA-N
4-3-chlorophenyl-1,7-diethylpyrido 2,3-d pyrimidin-2 1h-one,4-3-chlorophenyl-1,7-diethylpyrido 2,3-d pyrimidin-2-one,tocris-1821,d0c7jt,ym hplc,4-3-chlorophenyl-1,7-diethyl-1h,2h-pyrido 2,3-d pyrimidin-2-one,pyrido 2,3-d pyrimidin-2 1h-one,4-3-chlorophenyl-1,7-diethyl
6604918
4-(3-chlorophenyl)-1,7-diethylpyrido[2,3-d]pyrimidin-2-one
CCC1=NC2=C(C=C1)C(=NC(=O)N2CC)C3=CC(=CC=C3)Cl

PDE4 inhibitor

Orally active PDE4 inhibitor (IC 50 = 2.2 nM). Low emetogenic activity, suggested to be due to poor brain penetration.
Quantity 10mg
Formula Weight 313.79
Percent Purity >98%
Chemical Name or Material YM 976

Recommended Storage : Store at Room Temperature