Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Youngstown Barrel & Drum Ultra-Containment Trays
16.5 gallon sump is perfect for 5-gallon pails, batteries, safety cans, dry cleaning waste drums and leaky machine parts for spill containment.
Supplier: Youngstown Barrel & Drum 2328 UT
Description
Ultra-Containment Tray, No grate, black. 54 in. x 29¾ in. x 3½ in.. This low profile, 16.5 gallon sump is perfect for 5-gallon pails, batteries, safety cans, dry cleaning waste drums and leaky machine parts for spill containment. Polyethylene construction will not rust or corrode and provides excellent compatibility for most chemicals.Compliance
Meets EPA Container Storage Regulations for small packages with grating in place
Specifications
Black | |
62.45 L | |
Ultra-Containment Tray | |
Tray only | |
Resists UV rays, rust, corrosion and most chemicals | |
54 x 29.75 x 3.5 in. (137.16 x 75.56 x 8.89 cm) | |
137.16 cm | |
75.56 cm | |
8.89 cm | |
8.89 cm | |
7.5 kg |
16.5 gal. | |
Polyethylene | |
Used in maintenance and repair operations, accommodates many small containers | |
1 Ea. | |
Rectangular | |
54 in. | |
29.75 in. | |
3.5 in. | |
3.5 in. | |
17 lb. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction