Youngstown Barrel & Drum Ultra-Spill Pallets Plus™ Available on GSA/VA Contract for Federal Government customers only.

Low-profile design—only 8.75 in. high (22.2cm)

Manufacturer: Youngstown Barrel & Drum Company 9631 UT

 View more versions of this product

Catalog No. 19-130-4780

  Check Availability
Add to Cart

    Specifications

    Load Bearing Capacity (Metric) 4082kg
    Width (English) 62 in.
    Material Polyethylene
    Capacity (English) 75 gal.
    Capacity (Metric) 283.9L
    Capacity 75 gal., 283.9L
    Certifications/Compliance SPCC, EPA, 40 CFR 264.175, UFC
    Color Yellow
    Dimensions (L x W x H) 62 x 62 x 8.75 in. (157.4 x 157.4 x 22.22cm)
    For Use With (Application) Reduces the potential for hazardous fluids to splash into the eyes or face
    Height (English) 8.75 in.
    Height (Metric) 22.22cm
    Holds 4 Drums
    Includes Drain
    Length (English) 62 in.
    Length (Metric) 157.4cm
    Load Bearing Capacity (English) 9000 lb.
    Model P4
    Weight (English) 140 lb.
    Weight (Metric) 63.5kg
    Width (Metric) 157.4cm
    Product Type With drain

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit