Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Storage Bags for Ultra-Pop Up Pools™ Available on GSA/VA Contract for Federal Government customers only.

For use with Ultra-Pop Up Pools (cat. no. 19058085)

Manufacturer:  Youngstown Barrel & Drum Company 8158 UT

 View more versions of this product

Catalog No. 19-130-4809


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Yellow
66 gal. economy Ultra-pop™ up pool
3 lb.
Polyethylene
Used as decontamination pool and collection pool for soiled sorbents
Chemical, diesel fuel, antifreeze, acids, caustics and corrosives
1.3kg
Storage Bag
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit