Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Youngstown Barrel & Drum Storage Bags for Ultra-Pop Up Pools™
For use with Ultra-Pop Up Pools (cat. no. 19058085)
Supplier: Youngstown Barrel & Drum 8156 UT
Description
Order Info
Contact your Fisher Safety Customer Service Representative for ordering information.
Specifications
Yellow | |
Storage Bag | |
100 and 150 gal. economy Ultra-pop™ up pool | |
3 lb. |
Polyethylene | |
Used as decontamination pool and collection pool for soiled sorbents | |
Chemical, diesel fuel, antifreeze, acids, caustics and corrosives | |
1.3 kg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction