Learn More
Youngstown Barrel & Drum Ultra-Spill Berm™ Available on GSA/VA Contract for Federal Government customers only.
Helps contain or divert larger volume spills
Manufacturer:  Youngstown Barrel & Drum Company 2052 UT
 View more versions of this product
Catalog No. 19-130-4975
  Check Availability
Add to cart
Specifications
Description
Use for spill response, temporary secondary containment, and sealing off doorways during washdown operations
  • Flexible polyurethane construction is nonabsorbent and chemical resistant
  • Easily cleans with soap and water for repeated use
  • Spill Berm requires connectors and corners to connect 10 ft. sections and to make 90° corners (sold separately)
  • Spill Berm Plus features built-in connectors allowing multiple 5 ft. units to be quickly and easily connected
Description & Specifications
Specifications
NPDES, SPCC
10 x 2.25 x 1.37 in. (3.04m x 5.7 x 3.47cm)
1.37 in.
Self-sealing interlocking end joints
3.04m
5.9kg
5.7cm
Low Profile Ultra-Spill Berm™
Red
Use for spill response, temporary secondary containment, and sealing off doorways during washdown operations
3.47cm
10 ft.
13 lb.
2.25 in.
Polyurethane