Youngstown Barrel & Drum Ultra-Gravel Bags™ Available on GSA/VA Contract for Federal Government customers only.

Woven material eliminates the flaws and failures of sewn edges

Manufacturer: Youngstown Barrel & Drum Company 9782 UT

 View more versions of this product

Catalog No. 19-171-324

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) For drain protection and erosion control
    Product Type Ultra Gravel Bag
    Material Polymer
    Weight (English) 60 lb.
    Weight (Metric) 27.21kg
    Packaging Quantity 250 Bags/Box
    Color Green

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit