Learn More
Youngstown Barrel & Drum Ultra-Pop Up Pools Available on GSA/VA Contract for Federal Government customers only.
Instantly expand for quick response to emergency spills
Manufacturer:  Youngstown Barrel & Drum Company 8022 UT
 View more versions of this product
Catalog No. 19-820-945
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
20 gal.
20 gal., 75.7L
Used as decontamination pool and collection pool for soiled sorbents
Economy
10mil
Economy Ultra-Pop Up Pool™
75.7L
8 × 28 × 35 in. (20.3 × 71.1 × 89cm)
Storage bags
Chemical
Polyethylene