Youngstown Barrel & Drum Ultra-Microbes™ Available on GSA/VA Contract for Federal Government customers only.

“Supercharge” mother nature's own bioremediation process

Manufacturer: Youngstown Barrel & Drum Company 5239 UT

 View more versions of this product

Catalog No. 18-938-416

  Check Availability
Add to Cart

    Specifications

    Capacity (English) 28 oz.
    Capacity (Metric) 0.82L
    Capacity 28 oz., 0.82L
    For Use With (Application) For moist environment
    Quantity Each
    Shelf Life 4 Years
    Product Type Microbes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit