Learn More
Ultra-Concrete Washout Berm
Light-weight, portable way to collect harmful industrial concrete sedimant, drywall, mud, stucco and mortar from your work site.
Manufacturer:  Youngstown Barrel & Drum Company 3505 UT
 View more versions of this product
Catalog No. 18-938-422
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
NPDES, 40 CFR 122.26
Designed to replace filtering pods located in the third chamber of a concrete Washout Filter™, used in combination with a neutralization packets to help pull sediment out of suspension and reduce pH levels
Ultra-concrete washout filter™
Replacement Filter Pod