Youngstown Barrel & Drum Ultra-Containment Berms™, Collapsible Wall Model Available on GSA/VA Contract for Federal Government customers only.

Economical design offers cost savings and secure containment

Manufacturer: Youngstown Barrel & Drum Company 8400UT

 View all options for this product

Catalog No. 19-058-021

  Check Availability
Add to Cart


    Capacity (English) 748 gal.
    Capacity (Metric) 2831L
    Certifications/Compliance 40 CFR 264.175, SPCC, EPA
    Color Black
    Material Copolymer 2000™ an Ethylene Copolymer, Copolymer 3000™, Polyurethane, XR5, PVC
    Product Type Collapsible Wall Ultra-Containment Berm™
    Weight (English) 70 lb.
    Weight (Metric) 32kg
    Capacity 748 gal., 2831L

    Rugged design easily contains small and large spills

    • Heavy-duty copolymer 2000 construction with durable PVC sidewall assemblies for sidewall support
    • Raise or lower the sidewalls in seconds; simply swivel the feet of the PVC supports—vehicles and equipment can then roll over top the collapsed wall for convenient entry and exit
    • Fold up easily for compact storage and transport
    • Height: 12 in. (30.5cm)

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit