Youngstown Barrel & Drum Ultra-Containment Berms™, Compact and Stake Wall Models Available on GSA/VA Contract for Federal Government customers only.

Compact design folds into a tight package

Manufacturer: Youngstown Barrel & Drum Company 8610 UT

 View more versions of this product

Catalog No. 19-120-104

  Check Availability
Add to Cart

    Specifications

    Capacity (English) 269 gal.
    Capacity (Metric) 1018L
    Capacity 269 gal., 1018L
    Certifications/Compliance 40 CFR 264.175, SPCC, EPA
    For Use With (Application) Protects heavy equipment from spills during filling, discharging, cleaning and maintenance
    Weight (English) 17.5 lb.
    Weight (Metric) 39kg
    Material Copolymer 2000™
    Product Type Compact Ultra-Containment Berm™

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit