Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Containment Berm Ground Tarps

Place under berm to protect it from hazards that may damage it.

$1582.00 - $2110.00

Specifications

Color Black
Material Polyester Blended PVC
Product Type Ground Tarp
For Use With (Application) Protect the berm materials from punctures, tears and cuts that might occur from stones, glass, and other debris
For Use With (Equipment) For all 15 ft. x 50 ft. Containment Berms
View More Specs

Products 2
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity & Availability  
19-120-091
View Documents
Youngstown Barrel & Drum
8334 UT
70 ft. 21.3m Each for $2,110.00
Add to cart
 
19-120-093
View Documents
Youngstown Barrel & Drum
8338 UT
54 ft. 16.45m Each for $1,582.00
Add to cart
 
Description

Description

Containment Berm Ground Tarp. 19' x 54' For all 15' x 50' Containment BermsThis item go underneath the Berm and are typically 1 to 4 feet larger than the Berm itself. It is made from a 18.5 oz/sq. yd. polyester blended PVC.
Specifications

Specifications

Black
Ground Tarp
For all 15 ft. x 50 ft. Containment Berms
5.8m
Polyester Blended PVC
Protect the berm materials from punctures, tears and cuts that might occur from stones, glass, and other debris
19 ft.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit