Youngstown Barrel & Drum Open Head Steel Drum Available on GSA/VA Contract for Federal Government customers only.

Youngstown Open Head Steel Drum is designed for the transport of hazardous solids or liquids.

Manufacturer: Youngstown Barrel & Drum 392

 View more versions of this product

Catalog No. 19-130-4688

  Check Availability
Add to Cart

    Specifications

    Freight Class 50
    Weight (Metric) 5.44kg
    Packaging Quantity Each
    Certifications/Compliance UN 1A2/Y1.4/100/Y42/S-DOT/UN
    Diameter (English) Inner 13.78 in.
    Diameter (Metric) Inner 35.01cm
    Gauge 20
    Head Style Open
    Height (English) 14 in.
    Height (Metric) 35.56cm
    Material Carbon Steel
    Weight (English) 12 lb.
    Capacity (Metric) 30.28L
    Color Yellow
    For Use With (Application) For transport of hazardous solids or liquids, Designed to overpack and ship damaged and/or leaking drums
    Product Type Unlined Drum

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit